missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NSF Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 21172-1-AP-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Required for vesicle-mediated transport. Catalyzes the fusion of transport vesicles within the Golgi cisternae. Is also required for transport from the endoplasmic reticulum to the Golgi stack. Seem to function as a fusion protein required for the delivery of cargo proteins to all compartments of the Golgi stack independent of vesicle origin.Spezifikation
NSF | |
Polyclonal | |
Unconjugated | |
NSF | |
NEM sensitive fusion protein, NSF, SKD2, Vesicle fusing ATPase, Vesicular fusion protein NSF | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18195, 4905 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.38 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P46459, P46460 | |
Nsf | |
NSF Fusion Protein Ag15422 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Produktvorschläge
Customers who viewed this item also viewed.
Viewing 1-5 of 12
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
NSF Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur