Learn More
NSF Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke:  Proteintech 21172-1-AP-20UL
 Lernen Sie weitere Varianten dieses
Artikelnummer. 16850785
  • 136.48 CHF / 20 Mikroliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
Required for vesicle-mediated transport. Catalyzes the fusion of transport vesicles within the Golgi cisternae. Is also required for transport from the endoplasmic reticulum to the Golgi stack. Seem to function as a fusion protein required for the delivery of cargo proteins to all compartments of the Golgi stack independent of vesicle origin.
Spezifikation
Spezifikation
NSF
Polyclonal
Unconjugated
NSF
NEM sensitive fusion protein, NSF, SKD2, Vesicle fusing ATPase, Vesicular fusion protein NSF
Rabbit
Antigen Affinity Chromatography
RUO
18195, 4905
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation
0.38 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P46459, P46460
Nsf
NSF Fusion Protein Ag15422
20 μL
Primary
Human, Mouse
Antibody
IgG
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen