missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NHEJ1 Mouse anti-Human, Clone: 1F8C7, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66552-1-IG-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Double-strand breaks in DNA result from genotoxic stresses and are among the most damaging of DNA lesions. This gene encodes a DNA repair factor essential for the nonhomologous end-joining pathway, which preferentially mediates repair of double-stranded breaks. Mutations in this gene cause different kinds of severe combined immunodeficiency disorders.Spezifikation
NHEJ1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9H9Q4 | |
NHEJ1 | |
NHEJ1 Fusion Protein Ag2496 | |
150 μL | |
Primary | |
Human | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1F8C7 | |
Unconjugated | |
NHEJ1 | |
NHEJ1, Protein cernunnos, XLF, XRCC4 like factor | |
Mouse | |
Protein A | |
RUO | |
79840 | |
Antibody | |
IgG2b |