missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NF-H Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 18934-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This antibody can recognize both NEFH and NEFM.
Involved in the maintece of neuronal caliber, neurofilaments are the intermediate filament proteins found specifically in neurons, and are composed predomitly of three major proteins called NF-L, NF-M and NF-H. Like most other intermediate filament proteins (IFPs), the expression of the different neuronal IFPs is both tissue-specific and developmentally regulated. NF-H has an important role in mature axons and is involved in amyotrophic lateral sclerosis (ALS).Spezifikation
NF-H | |
Polyclonal | |
Unconjugated | |
NEFH | |
NEFH, NEH, NF H, NF200 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
24587, 380684, 4744 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P12036, P16884, P19246 | |
Nefh | |
NF-H Fusion Protein Ag13517 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |