missing translation for 'onlineSavingsMsg'
Learn More

Neuroligin 2/NLGN2 Antibody, Novus Biologicals™

Artikelnummer. 18249943 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
100 μL
25 μL
Packungsgröße:
100 Mikroliter
25 Mikroliter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18249943 100 μL 100 Mikroliter
18608917 25 μL 25 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 18249943 Lieferant Novus Biologicals Lieferanten-Nr. NBP256665

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Polyclonal Antibody

Neuroligin 2/NLGN2 Polyclonal specifically detects Neuroligin 2/NLGN2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen Neuroligin 2/NLGN2
Anwendungen Immunocytochemistry, Immunofluorescence
Klassifikation Polyclonal
Konjugat Unconjugated
Verdünnung Immunocytochemistry/Immunofluorescence 1-4 ug/ml
Zusammensetzung PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gen-Alias EC 3.1.1, EC 3.1.1.1, KIAA1366neuroligin-2, neuroligin 2
Gensymbole NLGN2
Wirtsspezies Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LGEERFPVVNTAYGRVRGVRRELNNEIL
Reinigungsverfahren Affinity Purified
Menge 100 μL
Regulatorischer Status RUO
Forschungsgebiet Cognition and Behavior
Primär oder sekundär Primary
Gen-ID (Entrez) 57555
Testspezifität Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Zielspezies Human
Inhalt und Lagerung Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.