Learn More
NEU2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 24523-1-AP-20UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The NEU2 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human NEU2. This antibody recognizes human, mouse, rat antigen. The NEU2 antibody has been validated for the following applications: WB, ELISA analysis.
Spezifikation
| NEU2 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 4759 | |
| IgG |
| Western Blot, ELISA | |
| 267 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |