missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEIL3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11621-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
DNA glycosylase which prefers single-stranded DNA (ssDNA), or partially ssDNA structures such as bubble and fork structures, to double-stranded DNA (dsDNA). In vitro, displays strong glycosylase activity towards the hydantoin lesions spiroiminodihydantoin (Sp) and guanidinohydantoin (Gh) in both ssDNA and dsDNA; also recognizes FapyA, FapyG, 5-OHU, 5-OHC, 5-OHMH, Tg and 8-oxoA lesions in ssDNA. No activity on 8-oxoG detected. Also shows weak DNA-(apurinic or apyrimidinic site) lyase activity. In vivo, appears to be the primary enzyme involved in removing Sp and Gh from ssDNA in neonatal tissues. Seems to be an important facilitator of cell proliferation in certain populations, for example neural stem/progenitor cells and tumor cells, suggesting a role in replication-associated DNA repair.Spezifikation
NEIL3 | |
Polyclonal | |
Unconjugated | |
NEIL3 | |
DNA glycosylase FPG2, DNA glycosylase/AP lyase Neil3, Endonuclease 8 like 3, Endonuclease VIII like 3, FGP2, FPG2, hFPG2, hNEI3, Nei like protein 3, NEI3, NEIL3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
234258, 55247 | |
-20°C | |
Liquid |
Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8K203, Q8TAT5 | |
NEIL3 | |
NEIL3 Fusion Protein Ag2200 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |