missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nectin-3/PVRL3 Antibody (CL15364) - Azide and BSA Free, Novus Biologicals™
Alle ansehen Bio Techne ProdukteBeschreibung
Nectin-3/PVRL3 Monoclonal antibody specifically detects Nectin-3/PVRL3 in Human,Mouse,Rat samples. It is validated for Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry
Specifications
Specifications
| Antigen | Nectin-3/PVRL3 |
| Anwendungen | Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry |
| Klassifikation | Monoclonal |
| Klon | CL15364 |
| Konzentration | LYOPH |
| Konjugat | Unconjugated |
| Verdünnung | Western Blot 1 μg/mL, Immunohistochemistry-Paraffin 1:500-1:1000 |
| Zusammensetzung | Lyophilized in 5% Trehalose. |
| Gen-Alias | CD113 antigen, CDw113FLJ90624, DKFZp566B0846, nectin 3, nectin-3, poliovirus receptor-related 3, poliovirus receptor-related protein 3, PPR3, PRR3CD113, PVRR3 |
| Wirtsspezies | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?