Learn More
Abnova™ NDUFB4 (Human) Recombinant Protein
Human NDUFB4 full-length ORF ( AAH00855.1, 1 a.a. - 129 a.a.) recombinant protein with GST-tag at N-terminal.
Marke: Abnova™ H00004710-P01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
- Sequence: MSFPKYKPSSLRTLPETLDPAEYNISPETRRAQAERLAIRAQLKREYLLQYNDPNRRGLIENPALLRWAYARTINVYPNFRPTPKNSLMGALCGFGPLIFIYYIIKTERDRKEKLIQEGKLDRTFHLSY
Spezifikation
AAH00855.1 | |
NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 4, 15kDa | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NDUFB4 | |
GST |
4710 | |
Wheat germ expression system | |
25 μg | |
B15, CI-B15, MGC5105 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |