missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYO9B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12432-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes a member of the myosin family of actin-based molecular motor heavy chain proteins. The protein has four IQ motifs located in the neck domain that bind calmodulin, which serves as a light chain. The protein complex has a single-headed structure and exhibits processive movement on actin filaments toward the minus-end. The protein also has rho-GTPase activity. Polymorphisms in this gene are associated with celiac disease and ulcerative colitis susceptibility. Multiple transcript variants encoding different isoforms have been found for this gene.Spezifikation
MYO9B | |
Polyclonal | |
Unconjugated | |
MYO9B | |
CELIAC4, MYO9B, MYO9B variant protein, myosin IXB, MYR5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17925, 25486, 4650 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q13459, Q63358, Q9QY06 | |
Myo9b | |
MYO9B Fusion Protein Ag3108 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |