Learn More
MYH6 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke:  Proteintech CL488-22281-100UL
 Lernen Sie weitere Varianten dieses
Artikelnummer. 18020726
  • 423.57 CHF / 100 Mikroliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
The MYH6 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human MYH6. This antibody recognizes human, mouse, rat antigen. The MYH6 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Spezifikation
Spezifikation
MYH6
1000 μg/mL
alpha MHC, ASD3, MYH6, MYHC, MyHC alpha, MYHCA, Myosin 6, Myosin heavy chain 6
100 μL
4624
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence
CoraLite Plus 488
Rabbit
Primary
Human, Mouse, Rat
150 kDa
IgG
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen