Learn More
MSP/MST1 Antibody (S04-1D6), Novus Biologicals™
Rabbit Monoclonal Antibody
Marke: Novus Biologicals NBP3-19812-100ul
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
MSP/MST1 Monoclonal antibody specifically detects MSP/MST1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Spezifikation
MSP/MST1 | |
Monoclonal | |
0.3 mg/mL | |
Western Blot, Immunohistochemistry 1:20-1:50, Immunohistochemistry-Paraffin | |
D3F15S2, EC 3.4.21, hepatocyte growth factor-like protein homolog, HGFLhepatocyte growth factor-like protein, macrophage stimulating 1 (hepatocyte growth factor-like), Macrophage stimulatory protein, Macrophage-stimulating protein, MSPDNF15S2, NF15S2 | |
A synthetic peptide of human MSP/MST1 (Uniprot # P26927) | |
100 μg | |
Protein Kinase | |
4485 | |
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
S04-1D6 | |
Unconjugated | |
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Berichtigung von Produktinhalten
Name des Produkts
MSP/MST1 Antibody (S04-1D6), Novus Biologicals™ > 100 μg; Unconjugated
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur