missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MS4A6A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11930-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
May be involved in signal transduction as a component of a multimeric receptor complex.Spezifikation
MS4A6A | |
Polyclonal | |
Unconjugated | |
MS4A6A | |
4SPAN3, 4SPAN3.2, CD20 antigen like 3, CD20L3, CDA01, MS4A6, MS4A6A, MST090, MSTP090 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
64231 | |
-20°C | |
Liquid |
Western Blot | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9H2W1 | |
MS4A6A | |
MS4A6A Fusion Protein Ag2464 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |