missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPL45 Mouse anti-Human, Mouse, Rat, Clone: 1B4B4, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66531-1-IG-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. Pseudogenes corresponding to this gene are found on chromosomes 2p and 17q.Spezifikation
MRPL45 | |
Monoclonal | |
0.9 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9BRJ2, Q9D0Q7 | |
Mrpl45 | |
MRPL45 Fusion Protein Ag8207 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
1B4B4 | |
Unconjugated | |
MRPL45 | |
L45mt, MRPL45, MRP-L45 | |
Mouse | |
Protein G | |
RUO | |
287656, 67036, 84311 | |
-20°C | |
Liquid |