missing translation for 'onlineSavingsMsg'
Learn More

MRPL45 Mouse anti-Human, Mouse, Rat, Clone: 1B4B4, Proteintech

Mouse Monoclonal Antibody

Marke:  Proteintech 66531-1-IG-150UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16838495

  • 381.34 CHF / 150 Mikroliter
Verfügbar ab: 17-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. Pseudogenes corresponding to this gene are found on chromosomes 2p and 17q.
Spezifikation

Spezifikation

MRPL45
Monoclonal
0.9 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9BRJ2, Q9D0Q7
Mrpl45
MRPL45 Fusion Protein Ag8207
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
1B4B4
Unconjugated
MRPL45
L45mt, MRPL45, MRP-L45
Mouse
Protein G
RUO
287656, 67036, 84311
-20°C
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote