missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse SR-AI/MSR Antibody, R&D Systems™
Goat Polyclonal Antibody
Marke: R&D Systems AF1797
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
SR-AI/MSR Polyclonal specifically detects SR-AI/MSR in Mouse samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.
Spezifikation
| SR-AI/MSR | |
| Polyclonal | |
| Unconjugated | |
| AAA39747 | |
| MSR1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse SR-AI/MSR Trp79-Ser454 Accession # AAA39747 | |
| 100 μg | |
| Primary | |
| Detects mouse SR-A1/MSR in direct ELISAs and Western blots. | |
| Mouse | |
| IgG |
| Western Blot, Blockade of Receptor-ligand Interaction | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction | |
| CD204, CD204 antigen, Macrophage acetylated LDL receptor I and II, macrophage scavenger receptor 1, macrophage scavenger receptor type III, MSR1, phSR1, phSR2, SCARA1, SCARA1macrophage scavenger receptor types I and II, Scavenger receptor class A member 1, scavenger receptor class A, member 1, SR-A, SRAI | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4481 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur