missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse SR-AI/MSR Antibody, R&D Systems™

Goat Polyclonal Antibody
281.00 CHF - 769.00 CHF
Spezifikation
| Antigen | SR-AI/MSR |
|---|---|
| Konzentration | LYOPH |
| Verdünnung | Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction |
| Anwendungen | Western Blot, Blockade of Receptor-ligand Interaction |
| Klassifikation | Polyclonal |
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
18788797
![]() |
R&D Systems
AF1797-SP |
25 μg |
281.00 CHF
25 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
|
18041512
|
R&D Systems
AF1797 |
100 μg |
769.00 CHF
100 Mikrogramm |
Log in om dit product te kopen Registreer vandaag om een webaccount aan te maken | |||||
Beschreibung
SR-AI/MSR Polyclonal specifically detects SR-AI/MSR in Mouse samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.Spezifikation
| SR-AI/MSR | |
| Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction | |
| Polyclonal | |
| Goat | |
| Mouse | |
| CD204, CD204 antigen, Macrophage acetylated LDL receptor I and II, macrophage scavenger receptor 1, macrophage scavenger receptor type III, MSR1, phSR1, phSR2, SCARA1, SCARA1macrophage scavenger receptor types I and II, Scavenger receptor class A member 1, scavenger receptor class A, member 1, SR-A, SRAI | |
| MSR1 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Blockade of Receptor-ligand Interaction | |
| Unconjugated | |
| RUO | |
| AAA39747 | |
| 4481 | |
| Mouse myeloma cell line NS0-derived recombinant mouse SR-AI/MSR Trp79-Ser454 Accession # AAA39747 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse SR-A1/MSR in direct ELISAs and Western blots. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts