Learn More
Mouse anti-Human IgG (H+L) Secondary Antibody, Alkaline Phosphatase, Novus Biologicals™
Artikelnummer. 18283394 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
1 mg
Packungsgröße:
1 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18283394 1 mg 1 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 18283394 Lieferant Novus Biologicals Lieferanten-Nr. NBP172683
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Mouse Polyclonal Antibody
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen IgG (H+L)
Anwendungen Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Klassifikation Polyclonal
Konjugat Alkaline Phosphatase (AP)
Verdünnung Western Blot 1:500 - 1:2500, ELISA 1:2000 - 1:10000, Immunohistochemistry 1:200-1:1000
Wirtsspezies Mouse
Immunogen Human IgG whole molecule
Reinigungsverfahren Affinity Purified
Menge 1 mg
Regulatorischer Status RUO
Forschungsgebiet Adaptive Immunity, Immunology
Primär oder sekundär Secondary
Testspezifität This product was prepared from monospecific polyclonal ascites by immunoaffinity chromatography using Human IgG coupled to agarose beads followed by solid phase adsorption(s) to remove any unwanted reactivities. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Mouse Serum, Human IgG and Human Serum.
Zielspezies Human
Inhalt und Lagerung Store at 4C. Do not freeze.
Isotype IgG