missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMACHC Mouse anti-Human, Clone: 4G10A11, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66609-1-IG-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The exact function of the protein encoded by this gene is not known, however, its C-terminal region shows similarity to TonB, a bacterial protein involved in energy transduction for cobalamin (vitamin B12) uptake. Hence, it is postulated that this protein may have a role in the binding and intracellular trafficking of cobalamin. Mutations in this gene are associated with methylmalonic aciduria and homocystinuria type cblC.Spezifikation
MMACHC | |
Monoclonal | |
0.91 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9Y4U1 | |
MMACHC | |
MMACHC Fusion Protein Ag15001 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
4G10A11 | |
Unconjugated | |
MMACHC | |
cblC, MMACHC, RP11 291L19.3 | |
Mouse | |
Protein G | |
RUO | |
25974 | |
-20°C | |
Liquid |