missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMACHC Mouse anti-Human, Clone: 4G10A11, Proteintech
Mouse Monoclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | MMACHC |
---|---|
Klon | 4G10A11 |
Konzentration | 0.91 mg/mL |
Anwendungen | Western Blot |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16848645
|
Proteintech
66609-1-IG-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 07-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16838645
|
Proteintech
66609-1-IG-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 07-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The exact function of the protein encoded by this gene is not known, however, its C-terminal region shows similarity to TonB, a bacterial protein involved in energy transduction for cobalamin (vitamin B12) uptake. Hence, it is postulated that this protein may have a role in the binding and intracellular trafficking of cobalamin. Mutations in this gene are associated with methylmalonic aciduria and homocystinuria type cblC.Spezifikation
MMACHC | |
0.91 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
cblC, MMACHC, RP11 291L19.3 | |
MMACHC | |
IgG1 | |
Protein G | |
Antibody |
4G10A11 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q9Y4U1 | |
25974 | |
MMACHC Fusion Protein Ag15001 | |
Primary | |
-20°C | |
MMACHC |