missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MKNK1 (MNK1) Human Protein
MKNK1 and MKNK2 are the Map kinase-interacting kinases.
Marke: Invitrogen™ PV6024
Additional Details : Gewicht : 0.01000kg
Beschreibung
The Map kinase-interacting kinases are activated by Map kinases, regulate protein synthesis by phosphroylating eIF4E on Ser209, and may contribute to tumorigenesis.Spezifikation
NP_945324 | |
66.6kDa | |
Kinase proteins are shipped on dry ice and should be stored at -80°C. | |
Full Length | |
MAPKAPK Kinase Family | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
1mg | |
RUO - research use only | |
MKNK1 (MNK1) | |
Human | |
Insect |