missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ MIS12 Overexpression Lysate

Artikelnummer. 18220144 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
0,1 mg
Unit Size:
0.10 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
18220144 0,1 mg 0.10 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18220144 Supplier Novus Biologicals™ Supplier No. NBL113115

om dit product te kopen Registreer vandaag om een webaccount aan te maken

This item is not returnable. View return policy
The MIS12 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Zelllinie The lysate was created in HEK293T cells, using plasmid ID RC2789 and based on accession number NM_2439. The protein contains a C-terminal DDK tag.
Zur Verwendung mit (Anwendung) Western Blot
Menge 0,1 mg
Lagerungspuffer RIPA buffer
Reinheit Protein
Testspezifität Homo sapiens MIS12, MIND kinetochore complex component, homolog (yeast) (MIS12), mRNA.
Inhalt und Lagerung -80°C. Avoid freeze-thaw cycles.
Protein State Native

Nur für Forschungszwecke

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.