missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MICA Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12619-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
MICA is the higly polymorphic MHC (HLA) class I chain-related gene A. The protein product is expressed on the cell surface, although unlike canonical class I molecules does not seem to associate with beta-2-microglobulin. It is thought that MICA functions as a stress-induced antigen that is broadly recognized by intestinal epithelial gamma delta T cells.Spezifikation
MICA | |
Polyclonal | |
Unconjugated | |
MICA | |
MIC A, MICA, PERB11.1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100507436 | |
-20°C | |
Liquid |
Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q29983 | |
MICA | |
MICA Fusion Protein Ag3298 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |