missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MFN1 Mouse anti-Human, Clone: 3F11C11, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66776-1-IG-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Mitofusin 1 (MFN1) and the related protein MFN2 are mitochondrial membrane GTPase proteins that play a central role in mitochondrial metabolism and may be associated with obesity and/or apoptosis processes. MFN1 and MFN2 form homotypic and heterotypic complexes and coordinately regulate mitochondrial fusion and are essential for embryonic development. When ectopically expressed, MFN1 inhibits the apoptosis-associated amino-terminal conformation change in the apoptotic protein Bax but not its mitochondrial translocation, indicating that MFN1 is involved in the regulating the activation of Bax on the outer mitochondrial membrane.Spezifikation
MFN1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8IWA4 | |
MFN1 | |
MFN1 Fusion Protein Ag4890 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Western Blot, Immunohistochemistry (Paraffin) | |
3F11C11 | |
Unconjugated | |
MFN1 | |
DKFZp762F247, FLJ20693, Fzo homolog, hfzo1, hfzo2, MFN1, mitofusin 1, Transmembrane GTPase MFN1 | |
Mouse | |
Protein A | |
RUO | |
55669 | |
-20°C | |
Liquid |