missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MFN1 Mouse anti-Human, Clone: 3F11C11, Proteintech
Mouse Monoclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | MFN1 |
---|---|
Klon | 3F11C11 |
Konzentration | 1 mg/mL |
Anwendungen | Western Blot, Immunohistochemistry (Paraffin) |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16848965
|
Proteintech
66776-1-IG-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 08-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16838965
|
Proteintech
66776-1-IG-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 08-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Mitofusin 1 (MFN1) and the related protein MFN2 are mitochondrial membrane GTPase proteins that play a central role in mitochondrial metabolism and may be associated with obesity and/or apoptosis processes. MFN1 and MFN2 form homotypic and heterotypic complexes and coordinately regulate mitochondrial fusion and are essential for embryonic development. When ectopically expressed, MFN1 inhibits the apoptosis-associated amino-terminal conformation change in the apoptotic protein Bax but not its mitochondrial translocation, indicating that MFN1 is involved in the regulating the activation of Bax on the outer mitochondrial membrane.Spezifikation
MFN1 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
DKFZp762F247, FLJ20693, Fzo homolog, hfzo1, hfzo2, MFN1, mitofusin 1, Transmembrane GTPase MFN1 | |
MFN1 | |
IgG2a | |
Protein A | |
Antibody |
3F11C11 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
Q8IWA4 | |
55669 | |
MFN1 Fusion Protein Ag4890 | |
Primary | |
-20°C | |
MFN1 |