missing translation for 'onlineSavingsMsg'
Learn More

MFN1 Mouse anti-Human, Clone: 3F11C11, Proteintech

Mouse Monoclonal Antibody

Marke:  Proteintech 66776-1-IG-150UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16838965

  • 381.34 CHF / 150 Mikroliter
Verfügbar ab: 17-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Mitofusin 1 (MFN1) and the related protein MFN2 are mitochondrial membrane GTPase proteins that play a central role in mitochondrial metabolism and may be associated with obesity and/or apoptosis processes. MFN1 and MFN2 form homotypic and heterotypic complexes and coordinately regulate mitochondrial fusion and are essential for embryonic development. When ectopically expressed, MFN1 inhibits the apoptosis-associated amino-terminal conformation change in the apoptotic protein Bax but not its mitochondrial translocation, indicating that MFN1 is involved in the regulating the activation of Bax on the outer mitochondrial membrane.
Spezifikation

Spezifikation

MFN1
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8IWA4
MFN1
MFN1 Fusion Protein Ag4890
150 μL
Primary
Human
Antibody
IgG2a
Immunohistochemistry (Paraffin), Western Blot
3F11C11
Unconjugated
MFN1
DKFZp762F247, FLJ20693, Fzo homolog, hfzo1, hfzo2, MFN1, mitofusin 1, Transmembrane GTPase MFN1
Mouse
Protein A
RUO
55669
-20°C
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote