missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MARCKS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20661-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
Myristoylated alanine-rich protein kinase C substrate (MARCKS), also designated 80K or 80K-L, has been identified as a major cellular substrate for protein kinase C. Human MARCKS is a 332 amino acid protein with a calculated molecular weight of 31.534 kDa; however, it has been shown to run at 80-87 kDa on Western blot. The plasma membrane bound protein dissociates from the membrane upon phosphorylation by various PKC isoforms. In NIH/3T3 fibroblasts, PKC alpha and PKC epsilon, but not PKC delta, are responsible for MARCKS phosphorylation. MARCKS has been found to bind calmodulin, Actin and Synapsin and is a filamentous (F) Actin crosslinking protein.Spezifikation
MARCKS | |
Polyclonal | |
Unconjugated | |
MARCKS | |
80K L, 80K L protein, MACS, MARCKS, PKCSL, PRKCSL | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17118, 25603, 4082 | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P26645, P29966, P30009 | |
Marcks | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |