missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MARCKS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | MARCKS |
---|---|
Konzentration | 0.21 mg/mL |
Anwendungen | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16850235
|
Proteintech
20661-1-AP-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 15-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16840235
|
Proteintech
20661-1-AP-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 15-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Myristoylated alanine-rich protein kinase C substrate (MARCKS), also designated 80K or 80K-L, has been identified as a major cellular substrate for protein kinase C. Human MARCKS is a 332 amino acid protein with a calculated molecular weight of 31.534 kDa; however, it has been shown to run at 80-87 kDa on Western blot. The plasma membrane bound protein dissociates from the membrane upon phosphorylation by various PKC isoforms. In NIH/3T3 fibroblasts, PKC alpha and PKC epsilon, but not PKC delta, are responsible for MARCKS phosphorylation. MARCKS has been found to bind calmodulin, Actin and Synapsin and is a filamentous (F) Actin crosslinking protein.Spezifikation
MARCKS | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P26645, P29966, P30009 | |
17118, 25603, 4082 | |
Peptide | |
Primary | |
-20°C | |
MARCKS |
0.21 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
80K L, 80K L protein, MACS, MARCKS, PKCSL, PRKCSL | |
Marcks | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |