Learn More
Invitrogen™ Human MAP3K14 (NIK), GST Tag Recombinant Protein
Human MAP3K14 (NIK), GST Tag Recombinant Protein
Marke: Invitrogen™ PR8920A
Additional Details : Gewicht : 0.01000kg
Beschreibung
MAP3K14 phosphorylates and activates CHUK (IKKalpha) and IKBKB (IKK beta), which in turn phosphorylate IkB-alpha, releasing NF-KappaB leading to up-regulation of inflammatory response genes.
Spezifikation
NP_003945.2 | |
100μg | |
STE Unique Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris pH 7.5, 150 mM NaCl, 0.02% Trition X-100, 0.5 mM EDTA, 2 mM DTT, 50% Glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors | |
Liquid |
96.9 kDa | |
MAP3K14 (NIK) | |
Human | |
GST Tag | |
Ser⁄Thr Kinases: STE Group | |
Baculovirus | |
Recombinant, Catalytic | |
Signal Transduction |