missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAFB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20189-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene is a basic leucine zipper transcription factor that plays an important role in the regulation of lineage-specific hematopoiesis. The encoded nuclear protein represses ETS1-mediated transcription of erythroid-specific genes in myeloid cells. This gene contains no introns.Spezifikation
MAFB | |
Polyclonal | |
Unconjugated | |
MAFB | |
KRML, Maf B, MAFB, Transcription factor MafB | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16658, 54264, 9935 | |
-20°C | |
Liquid |
Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P54841, P54842, Q9Y5Q3 | |
Mafb | |
MAFB Fusion Protein Ag14102 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |