missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAFB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | MAFB |
---|---|
Konzentration | 0.4 mg/mL |
Anwendungen | Western Blot |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16839704
|
Proteintech
20189-1-AP-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 06-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16829704
|
Proteintech
20189-1-AP-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 06-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The protein encoded by this gene is a basic leucine zipper transcription factor that plays an important role in the regulation of lineage-specific hematopoiesis. The encoded nuclear protein represses ETS1-mediated transcription of erythroid-specific genes in myeloid cells. This gene contains no introns.Spezifikation
MAFB | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
P54841, P54842, Q9Y5Q3 | |
16658, 54264, 9935 | |
MAFB Fusion Protein Ag14102 | |
Primary | |
-20°C | |
MAFB |
0.4 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
KRML, Maf B, MAFB, Transcription factor MafB | |
Mafb | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |