Learn More
Ly-6G Rat anti-Mouse, FITC, Clone: 1A8, BD
Rat Monoclonal Antibody
Marke: BD Biosciences 551460
Additional Details : Gewicht : 0.00900kg
Beschreibung
The 1A8 antibody reacts with Ly-6G, a 21-25kDa GPI-anchored protein. In the bone marrow, Ly-6G is expressed on the majority of the largest cells, which are predominantly granulocytes, and not on lymphoid or erythroid cells. In the periphery, it is expressed on granulocytes. The mAb RB6-8C5 (Cat. No. 557445/553123) recognizes both Ly-6G and Ly-6C and blocks the binding of mAb 1A8 to Ly-6G. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Rat
Clone: 1A8
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: Ly-6G-transfected EL4J cell line
Flow Cytometry
Spezifikation
Ly-6G | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Ly-6G-transfected EL4J cell line | |
0.5 mg | |
Immunology | |
Mouse | |
IgG2a κ |
Flow Cytometry | |
1A8 | |
FITC | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.