Learn More
Ly-6G Rat anti-Mouse, FITC, Clone: 1A8, BD
Rat Monoclonal Antibody
Marke: BD Biosciences 561105
Additional Details : Gewicht : 0.01000kg
Beschreibung
The 1A8 monoclonal antibody specifically binds to Ly-6G, a 21-25-kDa glycosyolphosphatidylinositol-anchored protein. In the bone marrow, Ly-6G is expressed on the majority of the largest cells, which are predominantly granulocytes, and not on lymphoid or erythroid cells. In the periphery, it is expressed on granulocytes. The RB6-8C5 monoclonal antibody recognizes both Ly-6G and Ly-6C and blocks the binding of mAb 1A8 to Ly-6G.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Spezifikation
Ly-6G | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Ly-6G-transfected EL4J cell line | |
25 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
1A8 | |
FITC | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2a κ |
For Research Use Only.