missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRRC10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 21575-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
LRRC10 may play important roles in cardiac development and/or cardiac function.Spezifikation
LRRC10 | |
Polyclonal | |
Unconjugated | |
LRRC10 | |
HRLRRP, LRRC10 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
237560, 314848, 376132 | |
-20°C | |
Liquid |
Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5BKY1, Q8K3W2 | |
Lrrc10 | |
LRRC10 Fusion Protein Ag16117 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |