missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
LRP2 Monoclonal antibody specifically detects LRP2 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | LRP2 |
| Anwendungen | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Klassifikation | Monoclonal |
| Klon | PSH04-52 |
| Konzentration | 1 mg/mL |
| Konjugat | Unconjugated |
| Verdünnung | Western Blot 1:2000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:1000 |
| Zusammensetzung | PBS (pH7.4), 0.1% BSA and 40% Glycerol |
| Gen-Alias | calcium sensor protein, DBSlow-density lipoprotein receptor-related protein 2, EC 1.1.2.3, EC 3.4.21.9, Glycoprotein 330, GP330, Heymann nephritis antigen homolog, low density lipoprotein receptor-related protein 2, LRP-2, megalin |
| Wirtsspezies | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?