missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LRDD Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12119-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene contains a leucine-rich repeat and a death domain. This protein has been shown to interact with other death domain proteins, such as Fas (TNFRSF6)-associated via death domain (FADD) and MAP-kinase activating death domain.Spezifikation
LRDD | |
Polyclonal | |
Unconjugated | |
PIDD1 | |
LRDD, PIDD | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
293625, 55367, 57913 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9ERV7, Q9HB75 | |
PIDD1 | |
LRDD Fusion Protein Ag2761 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |