missing translation for 'onlineSavingsMsg'
Learn More

Loperamide hydrochloride, 98+%, Thermo Scientific™

142.12 CHF - 517.68 CHF

Chemische Identifikatoren

CAS 34552-83-5
Summenformel C29H34Cl2N2O2
Molekulargewicht (g/mol) 513.503
MDL-Nummer MFCD00058581
InChI-Schlüssel PGYPOBZJRVSMDS-UHFFFAOYSA-N
Synonym loperamide hydrochloride, loperamide hcl, imodium, suprasec, dissenten, fortasec, lopemid, lopemin, loperyl, imosec
PubChem CID 71420
ChEBI CHEBI:6533
IUPAC-Name 4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-N,N-dimethyl-2,2-diphenylbutanamide;hydrochloride
SMILES CN(C)C(=O)C(CCN1CCC(CC1)(C2=CC=C(C=C2)Cl)O)(C3=CC=CC=C3)C4=CC=CC=C4.Cl
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
15412989
Sicherheitsdatenblatt (SDS) Unterlagen ansehen
Thermo Scientific Alfa Aesar
J60168.06
5g
ONLINE EXCLUSIVE PRICE
155.00 CHF
142.12 CHF / 5 Gramm
Sparen 12.88 CHF
8% Rabatt
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
om dit product te kopen Registreer vandaag om een webaccount aan te maken
15422989
Sicherheitsdatenblatt (SDS) Unterlagen ansehen
Thermo Scientific Alfa Aesar
J60168.14
25g
ONLINE EXCLUSIVE PRICE
557.00 CHF
517.68 CHF / 25 Gramm
Sparen 39.32 CHF
7% Rabatt
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung

Beschreibung

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • At nanomolar concentrations, binds to mu-opioid receptors
  • L-type calcium channel blocker
Spezifikation

Chemische Identifikatoren

34552-83-5
513.503
PGYPOBZJRVSMDS-UHFFFAOYSA-N
71420
4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-N,N-dimethyl-2,2-diphenylbutanamide;hydrochloride
C29H34Cl2N2O2
MFCD00058581
loperamide hydrochloride, loperamide hcl, imodium, suprasec, dissenten, fortasec, lopemid, lopemin, loperyl, imosec
CHEBI:6533
CN(C)C(=O)C(CCN1CCC(CC1)(C2=CC=C(C=C2)Cl)O)(C3=CC=CC=C3)C4=CC=CC=C4.Cl

Spezifikation

Loperamide hydrochloride
C29H34Cl2N2O2
UN2811
loperamide hydrochloride, loperamide hcl, imodium, suprasec, dissenten, fortasec, lopemid, lopemin, loperyl, imosec
CN(C)C(=O)C(CCN1CCC(CC1)(C2=CC=C(C=C2)Cl)O)(C3=CC=CC=C3)C4=CC=CC=C4.Cl
513.503
CHEBI:6533
≥98%
34552-83-5
MFCD00058581
14,5571
PGYPOBZJRVSMDS-UHFFFAOYSA-N
4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-N,N-dimethyl-2,2-diphenylbutanamide;hydrochloride
71420
513.51
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P301+P310-P330-P501c

H301

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 252-082-4

missing translation for 'rtecsNumber' : TM4960000

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.