missing translation for 'onlineSavingsMsg'
Learn More

Lithium amide, 95%

Product Code. 11486840
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11486840

Brand: Thermo Scientific Alfa Aesar L16023.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7782-89-0 | H2LiN | 22.963 g/mol

Lithium amide is used in the preparation of active pharmaceutical ingredients and antioxidants. It acts as a catalyst for polymers, as nucleophiles and as strong bases. It serves as a reagent in the synthesis of antiinflamatory and preoresolving protectin D1, chemotype dipeptidyl peptidase IV inhibitors and sterically congested triarylamines. It finds application in dyes displaying large stokes shifts. In addition to this, it is used as a reagent for cross-coupling of aryl chlorides and amine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lithium amide is used in the preparation of active pharmaceutical ingredients and antioxidants. It acts as a catalyst for polymers, as nucleophiles and as strong bases. It serves as a reagent in the synthesis of antiinflamatory and preoresolving protectin D1, chemotype dipeptidyl peptidase IV inhibitors and sterically congested triarylamines. It finds application in dyes displaying large stokes shifts. In addition to this, it is used as a reagent for cross-coupling of aryl chlorides and amine.

Solubility
Slightly soluble in ethanol and liquid ammonia. Insoluble in anhydrous ether, benzene and toluene.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, acids, water and alcohols.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-89-0
Molecular Formula H2LiN
Molecular Weight (g/mol) 22.963
MDL Number MFCD00011093
InChI Key AFRJJFRNGGLMDW-UHFFFAOYSA-N
Synonym lithium amide, lithamide, lithium amide li nh2, lithiumamide, unii-7393omu9lk, lithium azanide, hsdb 649, linh2, lithium amide 25g, lithium amide, powder
PubChem CID 24532
IUPAC Name lithium;azanide
SMILES [Li+].[NH2-]

Specifications

Boiling Point 430°C
Melting Point 373°C
Quantity 100 g
Linear Formula LiNH2
UN Number UN1390
Merck Index 14,5522
Solubility Information Slightly soluble in ethanol and liquid ammonia. Insoluble in anhydrous ether,benzene and toluene.
Formula Weight 22.96
Percent Purity 95%
Sensitivity Air and moisture sensitive
Density 1.178
Chemical Name or Material Lithium amide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.