missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LHX6 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 21516-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes a member of a large protein family that contains the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator and may be involved in the control of differentiation and development of neural and lymphoid cells. Two alternatively spliced transcript variants encoding distinct isoforms have been described for this gene. Alternatively spliced transcript variants have been identified, but their biological validity has not been determined.Spezifikation
LHX6 | |
Polyclonal | |
Unconjugated | |
LHX6 | |
LHX6, LHX6.1, LIM homeobox 6, LIM homeobox protein 6, LIM/homeobox protein Lhx6, LIM/homeobox protein Lhx6.1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26468 | |
-20°C | |
Liquid |
Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9UPM6 | |
LHX6 | |
LHX6 Fusion Protein Ag16047 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |