missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LHX6 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | LHX6 |
---|---|
Konzentration | 0.23 mg/mL |
Anwendungen | Western Blot |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16851105
|
Proteintech
21516-1-AP-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16841105
|
Proteintech
21516-1-AP-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
This gene encodes a member of a large protein family that contains the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator and may be involved in the control of differentiation and development of neural and lymphoid cells. Two alternatively spliced transcript variants encoding distinct isoforms have been described for this gene. Alternatively spliced transcript variants have been identified, but their biological validity has not been determined.Spezifikation
LHX6 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9UPM6 | |
26468 | |
LHX6 Fusion Protein Ag16047 | |
Primary | |
-20°C | |
LHX6 |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
LHX6, LHX6.1, LIM homeobox 6, LIM homeobox protein 6, LIM/homeobox protein Lhx6, LIM/homeobox protein Lhx6.1 | |
LHX6 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |