missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LENG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11406-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The function of this protein remains unknown.Spezifikation
LENG1 | |
Polyclonal | |
Unconjugated | |
LENG1 | |
LENG1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
292535, 69757, 79165 | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96BZ8, Q9DB98 | |
Leng1 | |
LENG1 Fusion Protein Ag1924 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |