missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAT Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11326-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene is phosphorylated by ZAP-70/Syk protein tyrosine kinases following activation of the T-cell antigen receptor (TCR) signal transduction pathway. This transmembrane protein localizes to lipid rafts and acts as a docking site for SH2 domain-containing proteins. Upon phosphorylation, this protein recruits multiple adaptor proteins and downstream signaling molecules into multimolecular signaling complexes located near the site of TCR engagement. Alternative splicing results in multiple transcript variants encoding different isoforms.Spezifikation
LAT | |
Polyclonal | |
Unconjugated | |
LAT | |
LAT, LAT1, p36 38, pp36 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16797, 27040 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O43561, O54957 | |
LAT | |
LAT Fusion Protein Ag1868 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |