missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAT Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | LAT |
---|---|
Konzentration | 0.17 mg/mL |
Anwendungen | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16848323
|
Proteintech
11326-1-AP-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 08-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16838323
|
Proteintech
11326-1-AP-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 08-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The protein encoded by this gene is phosphorylated by ZAP-70/Syk protein tyrosine kinases following activation of the T-cell antigen receptor (TCR) signal transduction pathway. This transmembrane protein localizes to lipid rafts and acts as a docking site for SH2 domain-containing proteins. Upon phosphorylation, this protein recruits multiple adaptor proteins and downstream signaling molecules into multimolecular signaling complexes located near the site of TCR engagement. Alternative splicing results in multiple transcript variants encoding different isoforms.Spezifikation
LAT | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
O43561, O54957 | |
16797, 27040 | |
LAT Fusion Protein Ag1868 | |
Primary | |
-20°C | |
LAT |
0.17 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
LAT, LAT1, p36 38, pp36 | |
LAT | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |