missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRT34 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20449-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene is a member of the keratin gene family. As a type I hair keratin, it is an acidic protein which heterodimerizes with type II keratins to form hair and nails. The type I hair keratins are clustered in a region of chromosome 17q12-q21 and have the same direction of transcription.Spezifikation
KRT34 | |
Polyclonal | |
Unconjugated | |
KRT34 | |
Ha 4, HA4, Hair keratin, type I Ha4, hHa4, HKA4, K34, keratin 34, Keratin, type I cuticular Ha4, KRT34, KRTHA4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16672, 3885 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.54 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O76011, Q9D646 | |
KRT34 | |
KRT34 Fusion Protein Ag14295 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |