missing translation for 'onlineSavingsMsg'
Learn More

KIR2DL3/CD158b2 Antibody (MM0440-8P9), Alexa Fluor™ 750, Novus Biologicals™

Artikelnummer. 18096708 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
0.1 mL
Packungsgröße:
0.10 Milliliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18096708 0.1 mL 0.10 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 18096708 Lieferant Novus Biologicals Lieferanten-Nr. NBP211760AF750

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Mouse Monoclonal Antibody

KIR2DL3/CD158b2 Monoclonal specifically detects KIR2DL3/CD158b2 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen KIR2DL3/CD158b2
Anwendungen Flow Cytometry
Klassifikation Monoclonal
Klon MM0440-8P9
Konjugat Alexa Fluor 750
Verdünnung Flow Cytometry
Zusammensetzung 50 mM sodium borate with 0.05% sodium azide
Gen-Alias CD158 antigen-like family member B2, CD158b2 antigen, CD158B2CD158b, cl-6, killer cell immunoglobulin-like receptor 2DL3, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 3, Killer inhibitory receptor cl 2-3, KIR-023GB, KIRCL23, KIR-K7b, KIR-K7c, MGC129943, MHC class I NK cell receptor, natural killer associated transcript 2, natural killer cell inhibitory receptor KIR2DL3, Natural killer-associated transcript 2, NKAT, nkat2, NKAT-2, NKAT2a, NKAT2B, NKAT2GL183, NK-receptor, p58, p58 natural killer cell receptor clone CL-6, p58 NK receptor CL-6, p58.2 MHC class-I specific NK receptor, p58.2 MHC class-I-specific NK receptor
Gensymbole KIR2DL3
Wirtsspezies Mouse
Immunogen Human recombinant KIR2DL3/CD158b2
Reinigungsverfahren Protein G purified
Menge 0.1 mL
Forschungsgebiet Apoptosis
Primär oder sekundär Primary
Gen-ID (Entrez) 3804.0
Zielspezies Human
Inhalt und Lagerung Store at 4°C in the dark.
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.