missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIAA1602 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 21392-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
KIAA1602, also named as NCKAP5L, has 4 isoforms. The target band is about 139-145 kDa for phosphorylation.Spezifikation
KIAA1602 | |
Polyclonal | |
Unconjugated | |
NCKAP5L | |
FP1193, KIAA1602, Nck associated protein 5 like, NCKAP5L | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
315297, 380969, 57701 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6GQX2, Q9HCH0 | |
Nckap5l | |
KIAA1602 Fusion Protein Ag16050 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |