missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNQ1DN Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 21233-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Spezifikation
KCNQ1DN | |
Polyclonal | |
Unconjugated | |
KCNQ1DN | |
BWRT, HSA404617, KCNQ1 downstream neighbor, KCNQ1DN | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55539 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9H478 | |
KCNQ1DN | |
KCNQ1DN Fusion Protein Ag15703 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |