missing translation for 'onlineSavingsMsg'
Learn More

KAZALD1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 20815-1-AP-150UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16840405

  • 381.34 CHF / 150 Mikroliter
Verfügbar ab: 15-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

This gene encodes a secreted member of the insulin growth factor-binding protein (IGFBP) superfamily. It contains an N-terminal insulin growth factor-binding domain, a central Kazal-type serine protease inhibitor and follistatin-like domain, and a C-terminal immunoglobulin-like domain. Studies of the mouse ortholog suggest that this gene product may have a function in bone development and bone regeneration.
Spezifikation

Spezifikation

KAZALD1
Polyclonal
Unconjugated
KAZALD1
BONO1, FKSG28, FKSG40, IGFBP rP10, KAZALD1, UNQ2945/PRO21184
Rabbit
Antigen Affinity Chromatography
RUO
81621
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.31 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q96I82
KAZALD1
KAZALD1 Fusion Protein Ag14796
150 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote