missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Jagged 2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 19696-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The Notch signaling pathway is an intercellular signaling mechanism that is essential for proper embryonic development. Members of the Notch gene family encode transmembrane receptors that are critical for various cell fate decisions. JAG2 is one of several ligands that activate Notch and related receptors.Spezifikation
Jagged 2 | |
Polyclonal | |
Unconjugated | |
JAG2 | |
HJ2, JAG2, jagged 2, Jagged2, Protein jagged 2, SER2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3714 | |
-20°C | |
Liquid |
Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9Y219 | |
JAG2 | |
Peptide | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |