missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITM2A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 18306-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes a type II membrane protein that belongs to the ITM2 family. Studies in mouse suggest that it may be involved in osteo- and chondrogenic differentiation. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Spezifikation
ITM2A | |
Polyclonal | |
Unconjugated | |
ITM2A | |
BRICD2A, E25A, integral membrane protein 2A, ITM2A, Protein E25 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16431, 317218, 9452 | |
Antibody | |
IgG |
Western Blot, Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O43736, Q61500 | |
Itm2a | |
Peptide | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Liquid |